ContactUsLinkComponent
Stands for Imhoff sedimentation cones
Stands for Imhoff sedimentation cones
Catalog # VITL81056
Supplier:  VITLAB
undefined
Stands for Imhoff sedimentation cones
Catalog # VITL81056
Supplier:  VITLAB
Specifications
  • Description:
    W×D×H: 300×150×294 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Sedimentation racks, made of PMMA. Holds two Imhoff sedimentation cones. Base plate with depression for exact vertical positioning of the sedimentation cone.