Colour reference solutions, ready to use, according to Ph. Eur., Certipur®, Supelco®
Supplier: Merck
Certificates
About this item
Certipur® colour reference solutions are developed for ‘Method 1’ as described in chapter 2.2.2 of the European Pharmacopoeia.
- Stable solutions with a minimum shelf life of 3 years
- Ready to use solutions
- Certificate of Analysis included
Limited Time Sale
Select items below are currently on sale
Specifications
- M-Clarity quality level:MQ100