Pressure filter holders
About this item
These filter holders are ideal for ultra-cleaning or sterilising batch volumes of liquids.
- Filter holder has reservoir to hold liquid for batch filtration
- Autoclavable with membrane in place for sterile filtration
The cylindrical barrel holds 100 or 340 ml of liquid, which is filtered by externally applied gas pressure through a suitable filter supported on the holder base.
2 Options Available Below
Product Details & Documents
These filter holders are ideal for ultra-cleaning or sterilising batch volumes of liquids.
- Filter holder has reservoir to hold liquid for batch filtration
- Autoclavable with membrane in place for sterile filtration
The cylindrical barrel holds 100 or 340 ml of liquid, which is filtered by externally applied gas pressure through a suitable filter supported on the holder base.
Specifications for Product Family
Specifications
- Catalog No:
- MILFXX4004700
- MILFXX4004740
- Filtration area:
- 11,3 cm²
- 11,3 cm²
- Inlet/outlet connections:
- 9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
- 9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flask
- Max. inlet pressure:
- 6,9 bar
- 6,9 bar
Specifications
Product Family Options
Product Information
- MaterialCapacityFor ØHeightExt.ØAvailabilityPrice
- MILFXX4004700Pressure filter holderPressure filter holderStainless steel100 ml47 mm292 mm70 mmLog in for availability1 * 1 ST2 130,00 €Specifications:
More
Specifications
Filtration area11,3 cm²Cat. No.MILFXX4004700Inlet/outlet connections9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flaskMax. inlet pressure6,9 bar - MILFXX4004740Pressure filter holderPressure filter holderStainless steel340 ml47 mm533 mm70 mmLog in for availability1 * 1 ST2 460,00 €Specifications:
More
Specifications
Filtration area11,3 cm²Cat. No.MILFXX4004740Inlet/outlet connections9,5 mm hose connector/No. 8 perforated silicone stopper mounts in standard 1 l filtering flaskMax. inlet pressure6,9 bar
Accessories
Product Information
- Catalog #AvailabilityPrice
- Filter support screen, stainless steel, 47 mmFilter support screen, stainless steel, 47 mmMILFXX4004704Log in for availability1 * 1 ST - 230,00 €
Product Information
- Catalog #AvailabilityPrice
- Locking ring gasket, PTFE
Product Information
- Catalog #AvailabilityPrice
- Replacement parts kit, 47 mm holder includes 5 each top cap O-rings, locking ring gasket, filter sealing O-ring and screen gasketReplacement parts kit, 47 mm holder includes 5 each top cap O-rings, locking ring gasket, filter sealing O-ring and screen gasketMILFXX40047RKLog in for availability1 * 1 ST - 458,00 €
Product Information
- Catalog #AvailabilityPrice
- PVC tubing 3 m with I.Ø. 9,5 mm