Order Entry
Belgium
ContactUsLinkComponent
MultiSqueegee swivel with mono blade
MultiSqueegee swivel with mono blade
  129-0265
undefined
MultiSqueegee swivel with mono blade
  129-0265

 

  • Colour:
    Blue/white
  • Description:
    MultiSqueegee swivel
  • Length:
    50 cm
  • Pk:
    1

 

 

For dry or wet push and pull applications. An alternative to a broom in places where bristles are not permitted, e.g. healthcare floors and walls. The MultiSqueegee combines three components: an aluminum rail
, an elastomer rubber blade (washable at 70 °C
) and a universal polypropylene handle connector. The replaceable rubber blade has a single sharp edge that drags the liquid and finest dirt in one go.

  • All materials can be cleaned by washing at 70 °C
  • Components are lightweight and easy to dismantle and the blade can be replaced
  • Materials have been improved to provide greater resistance to wear and rust
  • Replaceable rubber blade has a single sharp edge that drags the liquid and finest dirt in one go
 : Please note that this rubber blade is delivered alone (without handle ref. VILE119966 which has to be ordered separately).