ContactUsLinkComponent
VWR® Q Path® Slide Box
VWR® Q Path® Slide Box
Catalog # 631-1079
Supplier:  VWR Collection
undefined
VWR® Q Path® Slide Box
Catalog # 631-1079
Supplier:  VWR Collection
Specifications
  • No. of slides:
    5
  • Colour:
    Translucent
  • Description:
    Transport packaging
  • Material:
    Plastic
  • W×D×H:
    82×30×18 mm
  • Pk:
    500
Specifications
About this item
This box for five slides prevents items from breaking or sticking. Can be used for applications in cytology, such as conventional smears.
W×D×H: 82×30×18 mm