Visitors coats, 4400
About this item
These disposable coats made of soft, breathable PP are designed to protect the wearer against non hazardous dusts and non hazardous light liquid splashes.
- Low-linting properties
- Breathable material to help reduce heat build-up
- Front zip closing (single way)
- Elasticated cuffs for a secure fit
- Internal pocket on the wearers left-hand side
Typical applications include: General maintenance, catering, electronics manufacturing, laboratories and hygiene applications in general.
Colour: White
Weight: 45 g/m²
7 Options Available Below
Product Details & Documents
These disposable coats made of soft, breathable PP are designed to protect the wearer against non hazardous dusts and non hazardous light liquid splashes.
- Low-linting properties
- Breathable material to help reduce heat build-up
- Front zip closing (single way)
- Elasticated cuffs for a secure fit
- Internal pocket on the wearers left-hand side
Typical applications include: General maintenance, catering, electronics manufacturing, laboratories and hygiene applications in general.
Colour: White
Weight: 45 g/m²
Specifications for Product Family
Specifications
- Catalog No:
- MMMA4400WS
- MMMA4400WM
- MMMA4400WL
- MMMA4400WXL
- MMMA4400W2XL
- MMMA4400W3XL
- MMMA4400W4XL
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Pockets:
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Laboratory|Food
- Laboratory|Food
- Laboratory|Food
- Laboratory|Food
- Laboratory|Food
- Laboratory|Food
- Laboratory|Food
- Weight:
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeHeightLengthChest sizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MMMA4400WSClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400WMClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400WLClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400WXLClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400W2XLClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400W3XLClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m² - Specifications:
More
Specifications
Cat. No.MMMA4400W4XLClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationLaboratory|FoodWeight45 g/m²