ContactUsLinkComponent
Sealent wipes, KIMTECH PREP*
Sealent wipes, KIMTECH PREP*
Catalog # 115-1556
Supplier:  KIMBERLY CLARK
undefined
Sealent wipes, KIMTECH PREP*
Catalog # 115-1556
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    500 sheets/pack
  • Colour:
    White
  • L×W:
    400×500 mm
  • Pk:
    500
Specifications
About this item
These soft and flexible wipes have a non woven open structure that is ideal for the removal of excess sealant prior to industrial painting or coating of a surface.
  • Textured open weave material traps and removes excess sealant
  • Pliable material helps remove sealant from hard to get to spaces
  • Low linting performance helps minimise the occurrence of defects and cross-contamination from particulate matter