Orders LinkContactUsLinkComponent
VWR® Q Path® MicroStar V Cassettes
VWR® Q Path® MicroStar V Cassettes
Supplier:   VWR Collection
VWR® Q Path® MicroStar V Cassettes
Supplier:  VWR Collection
Certificates
VWR® Q Path® MicroStar V Cassettes
VWR® Q Path® MicroStar V Cassettes
9 Options Available Below
Product Details & Documents
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded disposable lid, supplied in dispensers for 500 cassettes. Packed 3 boxes with 500 cassettes.
  • Easy to use cassette with small opening
  • Writing at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Red
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €
  • Pale purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 205,00 €
    205,00 €
    1 * 1.500 ST
       205,00 €