VWR®, Filter Cards for Cytology Centrifuges
Supplier: VWR Collection
Certificates
About this item
Cyto strips made of filter board for cytocentrifuges. Absorption of excess liquids from the staining of samples in cytologic analysis. Can be used in Shandon and Sakura Cyto-Tek® cytocentrifuges.
- For single and multiple chambers
Frequently Bought Together