Order Entry
Belgium
Orders LinkContactUsLinkComponent
Waste bin, B1, Tork®

Waste bin, B1, Tork®

Black, 50 L, 389×289×629 mm
Catalog # 115-1690
Supplier:   Essity
Waste bin, B1, Tork®
Catalog # 115-1690
Supplier:  Essity
Waste bin, B1, Tork®
Waste bin, B1, Tork®

Black, 50 L, 389×289×629 mm

Price
132,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Plastic.

  • Free standing or wall-mounted
  • Compliments other Tork Elevation dispensers
  • Completely hidden bin liner for a cleaner appearance

This high capacity bin is suitable for busy washrooms and washstations.

Specifications
  • Capacity:
    50 L
  • Colour:
    Black
  • W×D×H:
    389×289×629 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.