Ultra fine absorption grains
Supplier: asecos
Jump to:
About this item
Fine absorption granules for oil, alkalis, grease, motor fuels, varnishes.
- Officially tested and approved for use on traffic areas
- 0,3 to 0,7 mm granularity
- 10 kg sack binds up to 11 litres of heating oil
- Fills smallest cracks and openings
- Developed for special purposes where a minimum of waste and a maximum of absorption is needed
2 Options Available Below
Product Details & Documents
Fine absorption granules for oil, alkalis, grease, motor fuels, varnishes.
- Officially tested and approved for use on traffic areas
- 0,3 to 0,7 mm granularity
- 10 kg sack binds up to 11 litres of heating oil
- Fills smallest cracks and openings
- Developed for special purposes where a minimum of waste and a maximum of absorption is needed
Product Family Options
Product Information
- AbsorbencyPackedAvailabilityPrice
- MSGA14394Ultra fine absorption granules (0,3 - 0,7 mm)Ultra fine absorption granules (0,3 - 0,7 mm)11 l/sack10 kg/sackLog in for availability1 * 1 ST18,60 €
- MSGA18165Ultra fine absorption granules (0,3 - 0,7 mm)Ultra fine absorption granules (0,3 - 0,7 mm)11 l/sack3×10 kg/sackLog in for availability1 * 1 ST54,00 €