Orders LinkContactUsLinkComponent
Fine absorption grains
Fine absorption grains
Supplier:   asecos
Fine absorption grains
Supplier:  asecos
Certificates
Fine absorption grains
Fine absorption grains
2 Options Available Below
Product Details & Documents
Fine absorption granules for oil, alkalis, grease, motor fuels, varnishes officially tested and approved for use on traffic areas.
  • Granularity of 0,5 to 1,0 mm
  • 20 kg sack binds up to 18 litres of heating oil
  • Fills small cracks and openings
Highly recommended where a large contact with the surface and a quick absorption is needed.
Product Family Options
Product Information
  • Absorbency
    Packed
    Availability
    Price
  • MSGA9906
    Universal absorption granules (1B)
    Universal absorption granules (1B)
    18 l/sack
    20 kg/sack
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,00 €
    31,00 €
    1 * 1 ST
       31,00 €
  • MSGA18453
    Universal absorption granules (3B)
    Universal absorption granules (3B)
    18 l/sack
    3×20 kg/sack
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,80 €
    89,80 €
    1 * 1 ST
       89,80 €