Orders LinkContactUsLinkComponent
Universal absorbent rolls
Universal absorbent rolls
Supplier:   asecos
Universal absorbent rolls
Supplier:  asecos
Certificates
Universal absorbent rolls
4 Options Available Below
Product Details & Documents
Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Absorbency of a capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Standard material for small leaks and prevention
  • Heavyweight material for small leaks or the absortion of spillage
  • Rolls are perforated to take sufficient material as required
Product Family Options
Product Information
  • Packed
    L×W
    Availability
    Price
  • MSGA11314
    Standard universal roll
    Standard universal roll
    2 rolls/pack
    90 m×400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 246,00 €
    246,00 €
    1 * 1 ST
       246,00 €
  • MSGA11317
    Standard universal roll
    Standard universal roll
    1 roll/pack
    90 m×800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 246,00 €
    246,00 €
    1 * 1 ST
       246,00 €
  • MSGA9950
    Heavyweight universal roll
    Heavyweight universal roll
    1 roll/pack
    45 m×400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 235,00 €
    235,00 €
    1 * 1 ST
       235,00 €
  • MSGA9949
    Heavyweight universal roll
    Heavyweight universal roll
    2 rolls/pack
    45 m×800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 241,00 €
    241,00 €
    1 * 1 ST
       241,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Roll stand, wide with cutting edge for rolls up to 80 cm width
    Roll stand, wide with cutting edge for rolls up to 80 cm width
    MSGA9909
    Log in for availability
    1 * 1 ST - 244,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Roll stand, small, with cutting edge for rolls up to 40 cm width
    Roll stand, small, with cutting edge for rolls up to 40 cm width
    MSGA9908
    Log in for availability
    1 * 1 ST - 64,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm width
    Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm width
    MSGA9905
    Log in for availability
    1 * 1 ST - 69,00 €