Orders LinkContactUsLinkComponent
Labeling tapes, warning tapes
Labeling tapes, warning tapes
Supplier:   Brady
Labeling tapes, warning tapes
Supplier:  Brady
Certificates
Labeling tapes, warning tapes
Labeling tapes, warning tapes
3 Options Available Below
Product Details & Documents
Self-adhesive warning film for marking pillars, pylons, the edges of walls, gate arches and lifts.
  • Extremely durable
  • Waterproof, oil, alkali and acid resistant
  • Different colours available
Length: 33,0 m.
Product Family Options
Product Information
  • Colour
    Width
    Availability
    Price
  • BRDY255312
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Black/yellow
    50,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 27,40 €
    27,40 €
    1 * 1 ROU
       27,40 €
  • 116-0231
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Red/white
    75,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 51,40 €
    51,40 €
    1 * 1 ROU
       51,40 €
  • BRDY255313
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Warning tape, self-adhesive PVC, 0,138 mm thick
    Yellow/black
    75,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - 51,40 €
    51,40 €
    1 * 1 ROU
       51,40 €