Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
Supplier:   MURRAY S
Spray bottles
Supplier:  MURRAY S
Certificates
Spray bottles
2 Options Available Below
Product Details & Documents
HDPE, translucent.
Designed to allow comfortable grip and minimise fatigue. Adjustable from fine spray to single jet. Delivery 1,4 ml per stroke.
Product Family Options
Product Information
  • Capacity
    Height
    Availability
    Price
  • 500 ml
    206 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 471,00 €
    471,00 €
    1 * 100 ST
       471,00 €
  • 500 ml
    206 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,40 €
    9,40 €
    1 * 1 ST
       9,40 €