Orders LinkContactUsLinkComponent
Spigots
Supplier:   BURKLE
Spigots
Supplier:  BURKLE
Certificates
Spigots
4 Options Available Below
Product Details & Documents
Media-contacting parts PP, PE and PTFE.
  • Tight closing for complete seal
  • Easy to use
  • With connecting cap nut
The spigots fit all containers with R¾" connection threads. Screw thread adapters facilitate connection of spigots to nearly all containers. Pressure resistant up to 2 bar at 20 °C (without discharge bracket and grommet). With discharge angle and rotatable spout preventing drips. With tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm.
Not suitable for solidifying substances.
Product Family Options
Angled
Product Information
  • Thread
    Availability
    Price
  • 216-2854
    StopCock, angled
    StopCock, angled
    Inflowing side: 3/4" inner thread cap nut/outflowing side: spout/nozzle
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,00 €
    37,00 €
    1 * 1 ST
       37,00 €
Straight
Product Information
  • Thread
    Availability
    Price
  • 216-0940
    StopCock straight-through
    StopCock straight-through
    Inflowing side: 3/4" outer thread, 3/4" inner thread cap nut/outflowing side: 3/4" outer thread, 3/4" inner thread cap nut
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,40 €
    41,40 €
    1 * 1 ST
       41,40 €
  • 216-0941
    StopCock straight-through
    StopCock straight-through
    Inflowing side: 3/4" outer thread, 3/4" inner thread cap nut/outflowing side: 3/4" outer thread, 3/4" inner thread cap nut
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,80 €
    36,80 €
    1 * 1 ST
       36,80 €
  • 216-0942
    StopCock straight-through
    StopCock straight-through
    Inflowing side: 3/4" outer thread, 3/4" inner thread cap nut/outflowing side: 3/4" outer thread, 3/4" inner thread cap nut
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,80 €
    43,80 €
    1 * 1 ST
       43,80 €