Order Entry
Belgium
ContactUsLinkComponent
RESOURCE PHE Columns
RESOURCE PHE Columns
Catalog # 17-1186-01
Supplier:  Cytiva
undefined
RESOURCE PHE Columns
Catalog # 17-1186-01
Supplier:  Cytiva

Specifications

  • Flow rate:
    0,8 - 4,8 ml/min
  • Max. pressure:
    15 bar
  • Capacity:
    1 ml
  • Description:
    SOURCE 15 PHE (phenyl) column
  • Binding capacity:
    At least 25 mg albumin/ml
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Column hardware:
    PEEK
  • Bed dimensions:
    6,4×30 mm
  • Pk:
    1 ml

Specifications

About this item

RESOURCE PHE are prepacked SOURCE 15 PHE (phenyl) columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC). For fast, high-resolution purifications, RESOURCE columns are excellent choice for polishing and removal of trace impurities.

  • High performance at low back pressure, even with high sample load of up to 25 mg of protein
  • Designed for low- to medium- pressure systems ranging from a peristalic pump to ÄKTA design and FPLC systems

RESOURCE HIC columns are available with ether, isopropyl, or phenyl. Generally, RESOURCE PHE will have the strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively.

Delivery: Columns include connectors for high- and low-pressure chromatography equipment and instructions.