VWR® Traceable®, Highly Accurate Freezer Thermometers
These triple-display thermometers offer a choice of different probes. The bottle insulates the thermometer from rapid temperature changes when door is opened. The stainless steel handle probe (Ø×L: 3,2×300 mm) with 3 metre cable is waterproof and suitable for liquids, air/gas and semi-solids, with adjustable holder and VELCRO® tape.
- Maximum/minimum values on the last 24 hours (each hour) and the last 7 or 30 days (each day)
- Time and date stamp key: Time and date for all minimum and maximum readings
- Visual LED flashes and audible alarm sounds continuously even if temperature returns to non alarm range in 0,1 °C increments
Choice of four platinum probes with PTFE cable.
Calibration points:
620-1976 and 620-1977: −40 °C; 0 °C and 50 °C
620-1978: −80 °C; 0 °C; 50 °C and 190 °C
620-1979: −80 °C; −40 °C and 0 °C
: Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
: Supplied with two AAA batteries, PT probe, cable, bench stand, wall mount and serial numbered Traceable® certificate of calibration.
: Note: For longer-lasting performance of the cable, please insert the probe and cable through a freezer port. If sealing the door on the cable, please provide extra slack so that the cable is not pulled every time the door is opened and closed.
Frequently Bought Together