Hygen™ PULSE™ Mop System
About this item
HYGEN™ PULSE™ spray mop provides an ergonomic, bucket-less mopping system to help cover more square metres in less time than traditional systems.
- Patented onboard reservoir and solution system evenly dispenses three sprays of cleaner behind mop
- Unscrew the cap to manually refill or connect to the PULSE™ Caddy hose for automatic refills
- Holds up to 0,6 L of liguid solution to clean up to 80 m² of floor space
- Non-slip tip helps prevent the hanlde from sliding and falling when placed against a wall
The HYGEN™ PULSE™ high-capacity caddy allows users to easily clean more square meters, reducing the frequency of trips to the supply closet. The 7,5 L capacity will refill the Rubbermaid PULSE™ spray mop up to 12 times. The caddy is designed to fit in Rubbermaid cleaning carts for efficient on-the-go refills.
3 Options Available Below
Product Details & Documents
HYGEN™ PULSE™ spray mop provides an ergonomic, bucket-less mopping system to help cover more square metres in less time than traditional systems.
- Patented onboard reservoir and solution system evenly dispenses three sprays of cleaner behind mop
- Unscrew the cap to manually refill or connect to the PULSE™ Caddy hose for automatic refills
- Holds up to 0,6 L of liguid solution to clean up to 80 m² of floor space
- Non-slip tip helps prevent the hanlde from sliding and falling when placed against a wall
The HYGEN™ PULSE™ high-capacity caddy allows users to easily clean more square meters, reducing the frequency of trips to the supply closet. The 7,5 L capacity will refill the Rubbermaid PULSE™ spray mop up to 12 times. The caddy is designed to fit in Rubbermaid cleaning carts for efficient on-the-go refills.
Documents
TDS Pulse Mopping Kit
Datasheet
Product Family Options
Product Information
Microfibre wet mop pads
Product Information
- Catalog #ColourDimensionsDescriptionAvailabilityPrice
Disposable microfibre pads
Product Information
- Catalog #ColourDimensionsMaterialAvailabilityPrice
Microfibre dust pad
Product Information
- Catalog #ColourDimensionsDescriptionAvailabilityPrice
- Green444×142×15 mmHYGEN™ microfibre dust pad for dust, dirt, and debrisLog in for availability1 * 1 ST19,80 €