Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multimode columns, HiTrap™ Capto™ adhere

Multimode columns, HiTrap™ Capto™ adhere

Supplier:   Cytiva
Multimode columns, HiTrap™ Capto™ adhere
Supplier:  Cytiva
Certificates
Multimode columns, HiTrap™ Capto™ adhere

3 Options Available Below

Product Details & Documents

HiTrap Capto adhere is prepacked with Capto adhere, a multimodal BioProcess ion exchange resin designed to remove contaminats in post-protein A purification.

  • High capacity and productivity in flowthrough mode
  • Savings in time and operating costs with a two-step chromatographic process
  • Wide operational window of pH and conductivity
  • Prepacked HiTrap columns for easy screening and convenient process development

Capto adhere is a strong anion exchanger with multimodal functionality. The multimodal functionality gives a different selectivity compared to traditional anion exchangers. Capto adhere is designed for post-protein A purification of monoclonal antibodies.

Removal of leached protein A, aggregates, host cell proteins, nucleic acids, and viruses from monoclonal antibodies is performed in flowthrough mode at which the antibodies pass directly through the column while the contaminants are adsorbed. For optimal performance of Capto adhere, screening/optimization of loading conditions are required.

Specifications for Product Family

Specifications

  • Catalog No:
  • 29-4004-56
  • 28-4058-44
  • 28-4058-46
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol
  • 4...30 °C, 20% Ethanol

Specifications

Product Family Options

Product Information

  • Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Flow rate
    Availability
    Price
  • 29-4004-56
    HiTrap™ Capto™ adhere
    HiTrap™ Capto™ adhere
    PP
    75 µm
    7×25 mm
    1×1 ml
    <4 ml/min
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,00 €
    95,00 €
    1 * 1 ST
       95,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    29-4004-56
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4...30 °C, 20% Ethanol
  • 28-4058-44
    HiTrap™ Capto™ adhere
    HiTrap™ Capto™ adhere
    PP
    75 µm
    7×25 mm
    5×1 ml
    <4 ml/min
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 428,00 €
    428,00 €
    1 * 5 ST
       428,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    28-4058-44
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4...30 °C, 20% Ethanol
  • 28-4058-46
    HiTrap™ Capto™ adhere
    HiTrap™ Capto™ adhere
    PP
    75 µm
    16×25 mm
    5×5 ml
    <20 ml/min
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 010,00 €
    1 010,00 €
    1 * 5 ST
       1 010,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    28-4058-46
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4...30 °C, 20% Ethanol