ContactUsLinkComponent
Precast ExcelGel for SDS-PAGE
  17-1342-01
 :  Cytiva
undefined
Precast ExcelGel for SDS-PAGE
  17-1342-01
 :  Cytiva
  • Description:
    ExcelGel SDS buffer strips (anode and cathode)
  • Seq:
    01
  • Pk:
    1 SET
ExcelGel precast gels for SDS-PAGE are available as homogeneous or gradient gels. The homogeneous gels have 25 preformed sample wells for sample volumes from 5 to 17 µl. The gradient gels use sample application strips (26 samples up to 40 µl, 52 samples up to 20 µl), paper pieces or sample droplets. All ExcelGel SDS homogeneous and 8 - 18 gradient gels have a 33 mm stacking zone and a 77 mm separating zone.
  • Save time with ready-to-use, mechanically stable gels cast on plastic supports
  • Apply samples quickly and easily using preformed sample wells
  • Avoid making running buffer by using precast buffer strips
ExcelGel gradient gels are recommended for use with Immobiline DryStrip gels.