Orders LinkContactUsLinkComponent
Standing support, FIN
Standing support, FIN
Supplier:   bimos
Standing support, FIN
Supplier:  bimos
Certificates
Standing support, FIN
Standing support, FIN
6 Options Available Below
Product Details & Documents
The seat made from soft touch PU foam. Almost every employee has to work standing up for long periods of time. But standing over prolonged periods subjects the human body to significant stress and is responsible for a number of complaints relating to the heart and the circulatory and musculoskeletal systems.
  • Comfortable: extra-wide comfort seat
  • Stable base and robust aluminium column which is inclined by 4 degrees that ensure complete stability while moving and encourage the correct active working posture
  • Flexibility and freedom: the seat can be swivelled 20° to left and right
  • Easy to adjust with the handle underneath the seat
  • Space saving design
  • Light and easy to move around
Certifications: Corresponds to DIN 68 877 German GS "safety tested" mark of confirmity
Confines to quality management system DIN EN ISO 9001
Corresponds to regulation EWG Nr. 1836/93 and environmental management system DIN ISO 14001
Has a ANSI/BIFMA X5.1 certified test report 21207935_001 TUV Rheinland
Product Family Options
Product Information
  • Height
    Foot style
    With
    Availability
    Price
  • 132-1418
    Standing support, black seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, black seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    620 - 850 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
  • 132-1419
    Standing support, black seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, black seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    620 - 850 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
  • 132-0896
    Standing support, blue seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, blue seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    620 - 850 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
  • BIMO9144-2001-2234
    Standing support, blue seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, blue seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    660 - 930 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
  • 132-0897
    Standing support, grey seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, grey seat and blasted aluminium black frame, 8 kg, seat dimensions: 450×150×620/850 mm
    620 - 850 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €
  • BIMO9144-2002-2234
    Standing support, grey seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    Standing support, grey seat and blasted aluminium brilliant silver frame, 8 kg, seat dimensions: 450×150×620/850 mm
    620 - 850 mm
    Glides, changeable
    Gas-lift column
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 423,00 €
    423,00 €
    1 * 1 ST
       423,00 €