Order Entry
Belgium
ContactUsLinkComponent
RNA isolation, illustra™ RNAspin 96 Kit
RNA isolation, illustra™ RNAspin 96 Kit
Catalog # 25-0500-75
Supplier:  Cytiva
CAS Number:  
undefined
RNA isolation, illustra™ RNAspin 96 Kit
Catalog # 25-0500-75
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    illustra™ RNAspin 96 Isolation Kit, 4×96 Preps
  • Description:
    illustra™ RNAspin 96 Isolation Kit
  • Yield:
    20–100 µg

Specifications

About this item

illustra™ RNAspin 96 kit is a flexible, high-throughput solution for total RNA isolation.

  • Fast and efficient purification in a 96-well format
  • 96 samples can be parallel-processed with high reproducibility in under 70 minutes
  • Flexible processing by vacuum or centrifugation
  • Integrated wash plate eliminates risk of contamination
  • DNase I treatment removes genomic DNA, leading to high quality total RNA product

illustra™ RNAspin 96 isolation kit can be used to reproducibly isolate total RNA from cultured cells, tissue, bacteria or yeast. The isolated RNA is of a high enough quality and quantity (up to 100 μg) for multiple downstream experiments, including sensitive enzymatic applications like quantitative reverse transcription polymerase chain reaction (qRT-PCR), primer extension, and RNase protection assays.