Order Entry
Belgium
ContactUsLinkComponent
Reversed phase chromatography media, SOURCE™ 15RPC
Reversed phase chromatography media, SOURCE™ 15RPC
Catalog # 17-0727-20
Supplier:  Cytiva
CAS Number:  
undefined
Reversed phase chromatography media, SOURCE™ 15RPC
Catalog # 17-0727-20
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    ~15 µm
  • Pk:
    10 ml
  • Description:
    SOURCE™ 15RPC
  • Seq:
    01
  • pH range:
    1 - 12
  • Matrix:
    Polystyrene/divinylbenzene
  • Ligand:
    Matrix surface
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    ~10 mg BSA/ml resin at 300 cm/h, ~30 mg bacitracin/ml resin at 300 cm/h, ~50 mg insulin/ml resin at 300 cm/h
  • Type:
    Liquid

Specifications

About this item

SOURCE™ 15RPC is designed for reversed phase chromatography (RPC) with full pH range stability and suitable for polishing steps in industrial processes requiring high flow velocities and low back-pressure.

  • Mono-sized 15 µm rigid beads gives low back pressures and high flow rates allowing for high productivity
  • Designed for high resolution reversed phase chromatography of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • Bioprocess medium supported for industrial applications

A controlled pore size distribution and large specific surface area offer excellent capacity for a wide range of molecules, from small peptides and oligonucleotides up to large proteins. With full pH range stability, SOURCE™ 15RPC is a strong alternative to silica-based RPC matrices.

Available in a range of different bulk pack sizes for easy scale-up and process development.