Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

illustra™ NICK™ columns

Supplier:   Cytiva
illustra™ NICK™ columns
Supplier:  Cytiva
Certificates
illustra™ NICK™ columns

2 Options Available Below

Product Details & Documents

NICK™ columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labelled probe from unincorporated labelled nucleotides.

  • For rapid purification of labeled DNA (20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography
  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0,15% Kathon CG/ICP Biocide
  • Can accommodate sample volumes up to 100 μl

They are ready for immediate use, and operate by gravity flow. Tests show that at least 97% of radioactivity applied in a nick-translated mixture elutes in two well-separated peaks, corresponding to labeled DNA and unincorporated nucleotides, respectively. At least 90% of the applied DNA is recovered.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 17-0855-01
  • 17-0855-02
  • Description:
  • Illustra™ NICK™ columns
  • Illustra™ NICK™ columns
  • Target molecule:
  • DNA
  • DNA
  • Refine target molecule:
  • Amplified/modified DNA
  • Amplified/modified DNA
  • Sample type:
  • PCR and other enzymatic reactions
  • PCR and other enzymatic reactions
  • Method / Format:
  • Gel filtration
  • Gel filtration
  • Prep size:
  • Scalable
  • Scalable

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 17-0855-01
    Illustra™ NICK™ columns
    Illustra™ NICK™ columns
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 249,00 €
    249,00 €
    1 * 20 ST
       249,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-0855-01
    Description
    Illustra™ NICK™ columns
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    PCR and other enzymatic reactions
    Method / Format
    Gel filtration
    Prep size
    Scalable
  • 17-0855-02
    Illustra™ NICK™ columns
    Illustra™ NICK™ columns
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 542,00 €
    542,00 €
    1 * 50 ST
       542,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-0855-02
    Description
    Illustra™ NICK™ columns
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    PCR and other enzymatic reactions
    Method / Format
    Gel filtration
    Prep size
    Scalable