Order Entry
Belgium
ContactUsLinkComponent
Ion exchange chromatography media, DEAE Sepharose™ CL-6B
Ion exchange chromatography media, DEAE Sepharose™ CL-6B
Catalog # 17-0710-01
Supplier:  Cytiva
CAS Number:  
undefined
Ion exchange chromatography media, DEAE Sepharose™ CL-6B
Catalog # 17-0710-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    90 µm
  • Pk:
    500 mL
  • Description:
    DEAE Sepharose™ CL-6B
  • pH range:
    3 - 9
  • Matrix:
    6% Cross-linked agarose
  • Ligand:
    Diethylaminoethyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    Thyroglobulin (MW 669 000) 2 mg/ml, HSA (MW 68 000) 170 mg/ml, alpha-lactalbumin (MW 14 300) 150 mg/ml
  • Total ionic capacity:
    0,13 - 0,17 mmol/ml
  • Flow rate:
    <150 cm/h
  • Type:
    Liquid

Specifications

About this item

DEAE Sepharose™ CL-6B is a weak anion exchanger.

  • Excellent flow properties
  • High capacity for proteins of all pI values

The ion exchange group is diethylaminoethyl which remains charged and maintains consistently high capacities over the entire working range, pH 3 to 9. Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride.