Order Entry
Belgium
ContactUsLinkComponent
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media
Catalog # 17-0810-01
Supplier:  Cytiva
CAS Number:  
undefined
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media
Catalog # 17-0810-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    90 µm
  • Pk:
    200 mL
  • Description:
    Phenyl Sepharose™ CL-4B
  • pH range:
    3 - 12
  • Matrix:
    Cross-linked agarose, 4%, spherical
  • Ligand:
    Phenyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    HSA (Mr 68 000) 15-20 mg/ml drained resin, ß-lactoglobulin (Mr 18 000) 3-5 mg/ml drained resin
  • Ligand density:
    40 μmole/ml drained medium
  • Flow rate:
    150 cm/h
  • Type:
    Liquid

Specifications

About this item

Octyl-Sepharose™ CL-4B is a well-proven medium for hydrophobic interaction chromatography (HIC). Octyl immobilised on 4% cross-linked agarose.

  • Phenyl immobilized on 4% cross-linked agarose
  • Aromatic ligand
  • Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
  • Negligible volume variation due to changes in pH or ionic strength

Octyl Sepharose® CL-4B and Phenyl Sepharose® CL-4B are covalently coupled to a cross-linked 4% agarose matrix by ether linkage, resulting in hydrophobic gels with minimal leakage and no ionic properties.