Order Entry
Belgium
ContactUsLinkComponent
 
Affinity chromatography columns, His GraviTrap™
  11-0033-99
 :  Cytiva
undefined
Affinity chromatography columns, His GraviTrap™
  11-0033-99
 :  Cytiva

 

  • Particle Size:
    90 µm
  • Capacity:
    1 ml
  • Description:
    His GraviTrap™
  • Seq:
    01
  • pH range:
    3 - 12
  • Binding capacity:
    Approx. 40 mg histidine-tagged protein/column
  • Column hardware:
    PP
  • Pk:
    10

 

 

His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification.

  • For fast and simple purification of histidine-tagged proteins
  • Convenient, prepacked gravity-flow column
  • Allows direct purification of both clarified and unclarified cell lysates
  • High binding capacity; approximately 40 mg of pure protein per column
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives

The ready to use column is prepacked with Ni Sepharose 6 Fast Flow, which has a high protein binding capacity, low nickel-ion leakage, and is compatible with denaturing and reducing agents, as well a wide range of additives.

His GraviTrap columns are delivered in a package that can be converted into a column stand to simplify purification. LabMate PD-10 Buffer Reservoir can be connected to the columns for convenient handling of sample volumes above 10 ml.