Order Entry
Belgium
ContactUsLinkComponent
 
Hydrophobic interaction chromatography media, Butyl Sepharose™ 4 Fast Flow
 :  Cytiva
 
undefined
Hydrophobic interaction chromatography media, Butyl Sepharose™ 4 Fast Flow
 :  Cytiva

 

  • pH range:
    3 - 13
  • Matrix:
    Cross-linked agarose, 4%, spherical
  • Ligand:
    Butyl
  • Storage temperature:
    4 - 30 °C
  • Ligand density:
    ~40 µmol Butyl/ml
  • Flow rate:
    150 - 250 cm/h

 

Frequently Bought Together