Order Entry
Belgium
ContactUsLinkComponent
Substrate for Western blotting, ECF™
Substrate for Western blotting, ECF™
Catalog # RPN5785
Supplier:  Cytiva
CAS Number:  
Substrate for Western blotting, ECF™
Catalog # RPN5785
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    ECF™ substrate for Western blotting

Specifications

About this item

  • A fluorescent substrate for alkaline phosphatase-based detection of protein blots, for use with fluorescence scanning equipment such as Storm instruments (see Storm)
  • For use in western blotting, dot, and slot blotting applications
  • Sensitivity comparable to chemiluminescence
  • The wider dynamic range of fluorescence scanning systems compared with film allows visualization and quantitation of weak and dark bands in the same scan
  • Eliminates need for darkroom and film

Alkaline phosphatase dephosphorylates ECF Substrate to produce a fluorescent product, which localizes at the site of catalysis. Product fluoresces at 540 to 560 nm when excited at the appropriate wavelength.

Delivery: The following reagents are provided, sufficient for detecting 2500 cm² membrane: ECF Substrate, 60 ml, and dilution buffer.