Order Entry
Belgium
ContactUsLinkComponent
Desalting columns, HiPrep™
Desalting columns, HiPrep™
Supplier:  Cytiva
Certificates
undefined
Desalting columns, HiPrep™
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 17-5087-01
  • 17-5087-02
  • Max. pressure:
  • 1,5 bar
  • 1,5 bar
  • pH range:
  • 2 - 13
  • 2 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together