Order Entry
Belgium
ContactUsLinkComponent
Hydrophobic interaction chromatography media, Phenyl Sepharose™ High Performance
Hydrophobic interaction chromatography media, Phenyl Sepharose™ High Performance
Catalog # 17-1082-01
Supplier:  Cytiva
CAS Number:  
undefined
Hydrophobic interaction chromatography media, Phenyl Sepharose™ High Performance
Catalog # 17-1082-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    34 µm
  • Pk:
    75 ml
  • Description:
    Phenyl Sepharose™ High Performance
  • pH range:
    3 - 13
  • Matrix:
    Cross-linked agarose, spherical
  • Ligand:
    Phenyl
  • Storage temperature:
    4 - 30 °C
  • Type:
    Liquid

Specifications

About this item

Phenyl Sepharose™ High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium, designed for intermediate and polishing step purification steps when high resolution has priority.

  • Selectivity similar to Phenyl Sepharose™ 6 Fast Flow (low sub) but higher resolution due to the smaller 34 µm bead size
  • Bioprocess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding