Order Entry
Belgium
ContactUsLinkComponent
 
Gel filtration media, Sephadex™ LH-20
 :  Cytiva
 
undefined
Gel filtration media, Sephadex™ LH-20
 :  Cytiva
 

 

  • pH range:
    2 - 13
  • Matrix:
    Hydroxypropyl
  • Ligand:
    Cross-linked dextran
  • Storage temperature:
    4 - 30 °C
  • Flow rate:
    <60 cm/hour

 

Frequently Bought Together