Order Entry
Belgium
ContactUsLinkComponent
Affinity chromatography columns, HisTrap™ FF crude
Affinity chromatography columns, HisTrap™ FF crude
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HisTrap™ FF crude
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 11-0004-58
  • 29-0486-31
  • 17-5286-01
  • Binding capacity:
  • Approx. 40 mg histidine-tagged protein/mL medium
  • Approx. 40 mg histidine-tagged protein/mL medium
  • Approx. 40 mg histidine-tagged protein/mL medium
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together