Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Electrodes kit, 2-Needle Array™

 :   BTX
Electrodes kit, 2-Needle Array™
 :  BTX
 
Electrodes kit, 2-Needle Array™
Electrodes kit, 2-Needle Array™

 

 

These in vivo style electrode specifically designed for muscle gene delivery. Among the non-viral techniques for in vivo gene transfer, the direct injection of plasmid DNA into muscle is simple, inexpensive and safe. The electrode is available in two gap sizes, 5 mm and 10 mm.

  • Handle made of Delrin, needle made of stainless steel
  • 5 mm 2-Needle array and handle is designed for small muscle masses such as mouse tibialis
  • 10 mm 2-Needle array and handle is designed for larger muscle masses such as rat gastrocnemius

Generator compatibility: ECM 830 and ECM 2001

Voltage range: 0 to 500 V

Pulse length range: 10 µsec to 90 msec

Needle length: 20 mm

Handle length: 8 cm

 : Electrodes are supplied in a convenient kit that includes one 2-Needle Array handle and six 2-Needle Arrays

Product Family Options

 

  • Description
    Application
     
     
  • Two needle array kit, 10 mm gap
    In vivo drug or gene delivery, muscle gene therapy
       
       
    1 * 1 ST
    1 * 1 ST - 684,00 €
    684,00 €
    1 * 1 ST
       684,00 €

  • Two needle array kit, 5 mm gap
    In vivo drug or gene delivery, muscle gene therapy
       
       
    1 * 1 ST
    1 * 1 ST - 624,00 €
    624,00 €
    1 * 1 ST
       624,00 €