Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flatpack chambers

Flatpack chambers

Flat pack chamber, 1,83 mm gap, 1,5 ml, 50
Catalog # 700-4813
Supplier:   BTX
Flatpack chambers
Catalog # 700-4813
Supplier:  BTX
CAS Number:  
Flatpack chambers
Flatpack chambers

Flat pack chamber, 1,83 mm gap, 1,5 ml, 50

CAS Number:  
Price
684,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Flatpack chambers are primarily used for prokaryotic applications (bacterial or yeast transformations), however they are also often used for high efficiency stem cell transfection.

  • The flow-through construction of the 0,56 mm gap has a volume capacity from 10 to 85 μl. This design provides the unique combination of small sample volumes with field strengths as high as 40 kV/cm.

The flatpack chamber with 1,83 mm gap has a three-ply solid sandwich construction of stainless steel and mylar plastic and holds a volume of 1,5 ml, ideal for stems cells.

Flatpack chambers are gamma sterilised in individual packages.

Delivery information: Supplied in sets of 50 and may be used in the BTX safety stand (available separately, please enquire).

Specifications

Specifications

  • Pk:
    50
  • Description:
    Flat pack chamber, 1,83 mm gap, 1,5 ml

Specifications