Sterivex™ Membrane Filter Units, with Millipore Express® (PES) Membrane
: Merck
The Sterivex™ filter unit has a compact design ideal for large volume filtration using positive pressure.
- Filters can fit virtually any workflow
- Filters dispense into nearly any storage container
The filter units work with syringes, peristaltic pumps or pressure vessels, thus eliminates the risk of contamination associated with decanting or pipetting. Filter-sterilise volumes up to 1000 ml or 2000 ml, using pressure driven filter units (capacity depends on the type of membrane used in the device).
- Catalog No:
- MILFSVGPB1010
- MILFSVGP01050
- MILFSVGP01015
- MILFSVGPL10RC
- Filters:
- Millipore Express® (PES), 0,22 µm, hydrophilic
- Millipore Express® (PES), 0,22 µm, hydrophilic
- Millipore Express® (PES), 0,22 µm, hydrophilic
- Millipore Express® (PES), 0,22 µm, hydrophilic
- Capacity:
- 2000 ml
- 2000 ml
- 2000 ml
- 2000 ml
- Filtration area:
- 10 cm²
- 10 cm²
- 10 cm²
- 10 cm²
- Max. temperature:
- 45 °C
- 45 °C
- 45 °C
- 45 °C
Frequently Bought Together