Orders LinkContactUsLinkComponent
Sample containers, straight, with screw cap, WHEATON®
Sample containers, straight, with screw cap, WHEATON®
Sample containers, straight, with screw cap, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Sample containers, straight, with screw cap, WHEATON®
Sample containers, straight, with screw cap, WHEATON®
9 Options Available Below
Product Details & Documents
Polystyrene clear, with metal screw cap.
  • Manufactured from FDA approved virgin polystyrene
  • Supplied sterile and for in vitro use only
  • CE marked in accordance with European Directive 98/79/EC
  • All containers are leak tested to British Standard BS5213:1975(1989)
  • All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Type
    Colour
    Material
    Availability
    Price
  • 100 ml
    No label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 193,00 €
    193,00 €
    1 * 200 ST
       193,00 €
  • 150 ml
    No label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 153,00 €
    153,00 €
    1 * 120 ST
       153,00 €
  • 150 ml
    Plain label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 156,00 €
    156,00 €
    1 * 120 ST
       156,00 €
  • 250 ml
    No label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 80,10 €
    80,10 €
    1 * 50 ST
       80,10 €
  • 250 ml
    Plain label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 82,20 €
    82,20 €
    1 * 50 ST
       82,20 €
  • 100 ml
    Plain label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 199,00 €
    199,00 €
    1 * 200 ST
       199,00 €
  • 100 ml
    Printed label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 199,00 €
    199,00 €
    1 * 200 ST
       199,00 €
  • 150 ml
    Printed label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 156,00 €
    156,00 €
    1 * 120 ST
       156,00 €
  • 250 ml
    Printed label, sterile
    Clear
    PS/metal
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 80,50 €
    80,50 €
    1 * 50 ST
       80,50 €