Orders LinkContactUsLinkComponent
Warning posts, Bradylink™
Warning posts, Bradylink™
Fournisseur:   Brady
Warning posts, Bradylink™
Fournisseur:  Brady
Certificates
Warning posts, Bradylink™
Warning posts, Bradylink™
2 Options Available Below
Product Details & Documents
Cordon off potentially hazardous areas both indoors and out with these portable posts and chains. The lightweight yet stable structure of these posts will last for years, whether intended for temporary or permanent use. The posts and chains are made from PU and are also safe to use in areas with electrical hazards. The colours do not fade, even after outdoor use. To provide greater stability, the hollow base can be filled with water or sand. The chain is securely attached to the post with a large C-shaped hook.
  • Non conducting
  • Integrated UV light filter
  • Rust-free
Delivery information: Supplied with post bases and tops. Chains not included.
Product Family Options
Product Information
  • Dimensions
    Disponibilité
    Prix
  • 116-0338
    Post, black
    Post, black
    75×1040 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 888,00 €
    888,00 €
    1 * 6 ST
       888,00 €
  • 116-0340
    Post, yellow
    Post, yellow
    75×1040 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 888,00 €
    888,00 €
    1 * 6 ST
       888,00 €