Handheld colorimeter, DR900
705-0744
: Hach
- Model:DR900
- Light source:LED
- Description:Multiparameter photometer
- Environmentally Preferable:
- Battery life:6 Months
- Display:Backlit graphical display (240×160 P)
- Interfaces:USB port for downloading and/or uploading data
- Optical design:Silicon photodiode detector
- Photometric accuracy:±0,005 A @ 1,0 A nominal
- Photometric range:0 − 2 A
- Wavelengths:420, 520, 560, 610 nm
- Weight:500 g
- W×D×H:87×47×236 mm
- Pk:1
The DR900 is a robust multiparameter configured for immediate use.
- Dustproof, waterproof and shockproof construction
- Easy-to-use menu-driven step-by-step analysis
- 90 pre-programmed methods
- High-quality reagents are available for your applications. They are formulated to ensure exceptional performance and deliver results
Specially designed for field applications.
For a list of popular test kits for the handheld colorimeter, DR900 instrument please see the COMPLEMENTARY PRODUCTS Tab and for a more comprehensive selection of test kits available from VWR®, please search the website using the search term ETK.
: IP 67 (protection against water and dust).
: Supplied with 4-AA alkaline batteries, multilingual instruction manual, 2 glass sample cells of 1" (25 mm) round cells with 10, 20, 25 ml marks, 2 sample cells 1 cm/10 ml cells, sample cell adapter and USB cable with mini USB connector.