PIG® Poly Drip Decks
Supplier: NEW PIG
Jump to:
About this item
These poly drip decks capture drips from containers and drums before they cause slip-and-fall hazards. These decks keep leaky drums and containers from creating puddles and slick spots on the floors.
- Low-profile 15 cm high design is easy to load or unload; pumps and funnels are easy to access
- Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
- Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
- Textured grating adds traction and lifts out for easy sump access
- Translucent white sidewalls allow easy visual inspection
Partly made from post-industrial recycled polyethtylene.
3 Options Available Below
Product Details & Documents
These poly drip decks capture drips from containers and drums before they cause slip-and-fall hazards. These decks keep leaky drums and containers from creating puddles and slick spots on the floors.
- Low-profile 15 cm high design is easy to load or unload; pumps and funnels are easy to access
- Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
- Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
- Textured grating adds traction and lifts out for easy sump access
- Translucent white sidewalls allow easy visual inspection
Partly made from post-industrial recycled polyethtylene.
Product Family Options
Product Information
- ColourWeightW×D×HAvailabilityPrice
- 121-0185Drip deck, 42 L sump capacity, 680 kg load capacityDrip deck, 42 L sump capacity, 680 kg load capacityWhite/black9,1 kg660×660×150 mmLog in for availability1 * 1 ST158,00 €
- 121-0186Drip deck, 83 L sump capacity, 1360 kg load capacityDrip deck, 83 L sump capacity, 1360 kg load capacityWhite/black18,2 kg1320×660×150 mmLog in for availability1 * 1 ST241,00 €
- 121-0187Drip deck, 167 L sump capacity, 2722 kg load capacityDrip deck, 167 L sump capacity, 2722 kg load capacityWhite/black31,8 kg1320×1320×150 mmLog in for availability1 * 1 ST391,00 €