Orders LinkContactUsLinkComponent
Automatic burettes, PTFE key, class AS, Schellbach
 :   SciLabware
Automatic burettes, PTFE key, class AS, Schellbach
Automatic burettes, PTFE key, class AS, Schellbach
Automatic burettes, PTFE key, class AS, Schellbach
Pyrex® borosilicate glass, class AS, with PTFE key stopcock of 1,5 mm bore.
  • Automatic zeroing
  • Schellbach stripe, with blue enamel graduations
Tube with vertical white stripe and central blue ribbon for accurate reading of the meniscus (where the blue ribbon splits into a wide and narrow band) and features a 29/32 Quickfit® cone.
 : Complies with ISO 385 and DIN 12700
 : Burettes and reservoir bottles should be ordered separately.
Product Family Options
  • Capacity
    Division
    Tolerance
  • 10 ml
    0,02 ml
    0,02 ± ml
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
  • 25 ml
    0,05 ml
    0,03 ± ml
    1 * 1 ST
    1 * 1 ST - 271,00 €
    271,00 €
    1 * 1 ST
       271,00 €
  • 50 ml
    0,10 ml
    0,05 ± ml
    1 * 1 ST
    1 * 1 ST - 284,00 €
    284,00 €
    1 * 1 ST
       284,00 €
Accessories
  •  
    Description
  • BIBB3297/14AS
    Blowing ball, rubber, for automatic burettes, Pellet pattern
    1 * 1 ST - 32,20 €
  •  
    Description
  • BIBB3297/12
    Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
    1 * 1 ST - 66,30 €