

Mitomycin C
Catalog # J63193.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:334.33 g/mol
Merck Index:14,6215
Melting Pt:360 °C
Storage Temperature:Refrigerator
EINECS:200-008-6
UN:2811
ADR:6.1,II
:GHS06
:GHS08
CAS Number:50-07-7
Formula:C₁₅H₁₈N₄O₅
MDL Number:MFCD00078109