Bacitracin
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Inhibits cell wall synthesis in Gram-positive bacteria.
- White to off-white powder
Soluble in water, methanol or ethanol
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
- Merck Index:14,934
- Melting Pt:222 °C
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
Inhibits cell wall synthesis in Gram-positive bacteria.
- White to off-white powder
Soluble in water, methanol or ethanol
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
- Merck Index:14,934
- Melting Pt:222 °C
- Storage Temperature:Freezer
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



