Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

α-Cyclodextrin ≥97%

α-Cyclodextrin ≥97%
Certificates
α-Cyclodextrin ≥97%
α-Cyclodextrin ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97+% CAS No.: 10016-20-3 Molecular Formula: C36H60O30 Formula Weight: 972.84 Physical Form: Powder Appearance: White Solubility: Soluble in water at 1%(w/v) Merck Reference: 14,2718 Optical Rotation: +136° (c=10 in water) MDL No.: MFCD00078207

Useful for selective precipitation of enantiomeric, positional or structural isomers

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Merck Index:14,2718
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Product Family Options

Product Information