Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, granules, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Sodium carbonate, granules, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Sodium carbonate, granules, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (Na₂CO₃) (dried basis)(by acidimetry):
    ≥99.5 %
  • Insoluble Matter:
    ≤0.01 %
  • Loss on Heating at 285°C:
    ≤1.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Silica (SiO₂):
    ≤0.005 %
  • Sulfur Compounds (as SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.03 %
  • Magnesium (Mg):
    ≤0.005 %
  • Potassium (K):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together