Order Entry
Belgium
ContactUsLinkComponent
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (KBr) (by Ag titrn):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    5.0 - 8.8
  • Bromate (BrO₃):
    ≤0.001 %
  • Iodate (IO₃):
    ≤0.001 %
  • Chloride (Cl):
    ≤0.2 %
  • Iodide (I):
    ≤0.001 %
  • Nitrogen Compounds (as N):
    ≤0.005 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Barium (Ba):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.002 %
  • Magnesium (Mg):
    ≤0.001 %
  • Sodium (Na):
    ≤0.02 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together