



p-Anisaldehyde solution in ethanol (contains Acetic acid, H₂SO₄) stain, for TLC
Supplier: TCI
Certificates
About this item
CAS Number:123-11-5
:
Formula:C₈H₈O₂
MDL Number:MFCD00003385
Merck Index:14,00663
Boiling Pt:140 °C (1013 hPa)
Melting Pt:0…2 °C
Density:1,12 g/cm³ (25 °C)
Flash Pt:110 °C
UN:2924
ADR:3,II
:136.15 g/mol
EINECS:204-602-6
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light red to Green clear liquid
- Color test:A yellow spot appears on 4-dimethylaminobenzaldehyde.
Specification Test Results
Frequently Bought Together