Order Entry
Belgium
ContactUsLinkComponent
Citric acid 20% (w/v) in aqueous solution
Corrosive
Citric acid 20% (w/v) in aqueous solution
Catalog # CMPLZ12984068.5L
CAS Number:  
undefined
Corrosive
Citric acid 20% (w/v) in aqueous solution
Catalog # CMPLZ12984068.5L
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :192.13 g/mol
EINECS:201-069-1
 :GHS05
Storage Temperature:Ambient
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669