

Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # 9018-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤ 0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:≤ 0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤ 190
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤ 0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤ 1.0 ppb
- Appearance:Passes Test
- Assay (CH₃CN) (by GC, corrected for water):≥ 99.8 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 1.0 ppm
- Titrable Acid (µeq/g):≤ 0.8
- Titrable Base (µeq/g):≤ 0.6
- Water (by KF, coulometric):≤ 10 ppm
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878